Gene Transcripts

Gene Symbol
Plce1
Entrez Gene ID
74055
Gene Name
phospholipase C, epsilon 1
Gene Aliases
4933403A21Rik, PLCepsilon, Plce, mKIAA1516
Chromosome Location
Chr. 19: 38481114 - 38785105 on Build GRCm38
UniGene ID
Mm.34031
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019588.2 NP_062534.2 32 7537
GenBank mRNA AB076247.1 - 32 7536
AK122521.1 - 26 4723
AK162236.1 - 8 1748

View Full Product Details