Gene Transcripts

Gene Symbol
Esm1
Entrez Gene ID
71690
Gene Name
endothelial cell-specific molecule 1
Gene Aliases
0610042H23Rik, AV004503, ESM-1
Chromosome Location
Chr. 13: 113209659 - 113218104 on Build GRCm38
UniGene ID
Mm.38929
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023612.3 NP_076101.1 3 1466
GenBank mRNA AK002913.1 - 3 1360
BC020038.1 - 3 1352 4018658
BC051919.1 - 3 1466 6403364

View Full Product Details