Gene Transcripts

Gene Symbol
Mxra8
Entrez Gene ID
74761
Gene Name
matrix-remodelling associated 8
Gene Aliases
1200013A08Rik, 1700095D18Rik, AI131686, Asp3, Dicam
Chromosome Location
Chr. 4: 155839680 - 155844102 on Build GRCm38
UniGene ID
Mm.159028
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024263.4 NP_077225.4 8 1361
GenBank mRNA AB040488.1 - 8 1281
AB052097.1 - 8 1239
AK004732.1 - 8 1244
AK195482.1 - 2 85
BC002140.1 - 3 358
BC026438.1 - 8 1283 4239626
EU849596.1 - 8 1199
EU849597.1 - 8 1190

View Full Product Details