Gene Transcripts
- Gene Symbol
- Wdr13
- Entrez Gene ID
- 73447
- Gene Name
- WD repeat domain 13
- Gene Aliases
- 1700060B08Rik, 5730411P10Rik, DXHXS7467e, W51679, mMg21
- Chromosome Location
- Chr. X: 8123301 - 8132858 on Build GRCm38
- UniGene ID
- Mm.284162
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290783.1 |
NP_001277712.1
|
2, 3 |
771 |
|
NM_026137.5 |
NP_080413.2
|
2, 3 |
805 |
|
NM_028542.1 |
NP_082818.1
|
2, 3 |
1057 |
|
GenBank mRNA |
AF220146.1 |
- |
1, 2 |
327 |
|
AF353243.1 |
- |
2, 3 |
451 |
|
AF513713.1 |
- |
2, 3 |
274 |
|
AK006837.1 |
- |
1, 2 |
293 |
|
AK017549.1 |
- |
2, 3 |
387 |
|
BC037502.1 |
- |
2, 3 |
422 |
|
BC057366.1 |
- |
2, 3 |
771 |
|
BC080853.1 |
- |
2, 3 |
720 |
|
BC108332.1 |
- |
2, 3 |
561 |
6493226 |
BC120553.1 |
- |
2, 3 |
308 |
40129476 |
BC125287.1 |
- |
2, 3 |
307 |
40129802 |
View Full Product Details