Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006346.2 | NP_006337.2 | 2 | 436 | |
XM_011534881.2 | XP_011533183.1 | 2 | 464 | ||
XM_011534882.2 | XP_011533184.1 | 2 | 464 | ||
XM_011534884.2 | XP_011533186.1 | 2 | 464 | ||
XM_011534886.2 | XP_011533188.1 | 2 | 464 | ||
XM_017020351.1 | XP_016875840.1 | 2 | 464 | ||
XM_017020352.1 | XP_016875841.1 | 2 | 390 | ||
XM_017020353.1 | XP_016875842.1 | 2 | 387 | ||
XM_017020354.1 | XP_016875843.1 | 2 | 387 | ||
GenBank mRNA | AF330046.1 | - | 2 | 478 | |
AK309828.1 | - | 2 | 393 | ||
AY370776.1 | - | 2 | 335 | ||
BC006009.1 | - | 2 | 389 | ||
BC008432.1 | - | 2 | 389 | ||
BC041806.1 | - | 2 | 388 | 5269898 | |
BC051911.1 | - | 2 | 366 | 6140514 | |
BX341357.2 | - | 2 | 436 |