Gene Transcripts
- Gene Symbol
- Telo2
- Entrez Gene ID
- 71718
- Gene Name
- TEL2, telomere maintenance 2, homolog (S. cerevisiae)
- Gene Aliases
- 1200003M09Rik, AI415602, Tel2, mKIAA0683
- Chromosome Location
- Chr. 17: 25099569 - 25115971 on Build GRCm38
- UniGene ID
- Mm.288702
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163661.1 |
NP_001157133.1
|
12, 13 |
2141 |
|
NM_027880.2 |
NP_082156.2
|
12, 13 |
2052 |
|
GenBank mRNA |
AK004582.1 |
- |
12, 13 |
1922 |
|
AK028658.1 |
- |
12, 13 |
1906 |
|
AK045321.1 |
- |
5, 6 |
1588 |
|
AK050773.1 |
- |
12, 13 |
1871 |
|
AK084830.1 |
- |
12, 13 |
1923 |
|
AK129189.1 |
- |
11, 12 |
2214 |
|
AK143827.1 |
- |
12, 13 |
2142 |
|
AK146963.1 |
- |
12, 13 |
1928 |
|
AK154727.1 |
- |
12, 13 |
1954 |
|
BC011077.1 |
- |
12, 13 |
1902 |
3984911 |
BC080713.1 |
- |
11, 12 |
2161 |
|
View Full Product Details