Gene Transcripts

Gene Symbol
Telo2
Entrez Gene ID
71718
Gene Name
TEL2, telomere maintenance 2, homolog (S. cerevisiae)
Gene Aliases
1200003M09Rik, AI415602, Tel2, mKIAA0683
Chromosome Location
Chr. 17: 25099569 - 25115971 on Build GRCm38
UniGene ID
Mm.288702
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163661.1 NP_001157133.1 21 3431
NM_027880.2 NP_082156.2 21 3342
GenBank mRNA AK004582.1 - 21 3212
AK028658.1 - 21 3196
AK050773.1 - 21 3162
AK129189.1 - 18 4336
AK143827.1 - 21 3432
AK146963.1 - 21 3218
AK154727.1 - 21 3244
BC011077.1 - 21 3192 3984911
BC080713.1 - 18 3259

View Full Product Details