Gene Transcripts
- Gene Symbol
- ITSN2
- Entrez Gene ID
- 50618
- Gene Name
- intersectin 2
- Gene Aliases
- PRO2015, SH3D1B, SH3P18, SWA, SWAP
- Chromosome Location
- Chr. 2: 24202866 - 24360528 on Build GRCh38
- UniGene ID
- Hs.432562
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006277.2 |
NP_006268.2
|
3, 4 |
375 |
|
NM_019595.3 |
NP_062541.3
|
3, 4 |
375 |
|
NM_147152.2 |
NP_671494.2
|
3, 4 |
375 |
|
GenBank mRNA |
AB033082.1 |
- |
3, 4 |
282 |
|
AF182198.1 |
- |
3, 4 |
354 |
|
AF182199.1 |
- |
3, 4 |
354 |
|
AF248540.1 |
- |
2, 3 |
156 |
|
BC020921.2 |
- |
2, 3 |
152 |
|
BC038963.1 |
- |
3, 4 |
366 |
|
BC146779.1 |
- |
3, 4 |
282 |
9007231 |
DA382952.1 |
- |
3 |
625 |
|
View Full Product Details