Gene Transcripts

Gene Symbol
Ppp1r14c
Entrez Gene ID
76142
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 14c
Gene Aliases
6330514J04Rik, KEPI
Chromosome Location
Chr. 10: 3366071 - 3464979 on Build GRCm38
UniGene ID
Mm.308126
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133485.2 NP_597844.1 3 916
GenBank mRNA AF407167.2 - 3 1013
AK085997.1 - 3 954
BC055717.1 - 3 523 6415732

View Full Product Details