Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023343.2 | NP_075832.1 | 5 | 485 | |
GenBank mRNA | AK003007.1 | - | 4 | 1458 | |
AK005408.1 | - | 5 | 508 | ||
BC026953.1 | - | 5 | 485 | 5133980 | |
BC027439.1 | - | 5 | 374 | 4164301 |