Gene Transcripts
- Gene Symbol
- Ndufb8
- Entrez Gene ID
- 67264
- Gene Name
- NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8
- Gene Aliases
- 2900010I05Rik, AI987932
- Chromosome Location
- Chr. 19: 44548576 - 44555446 on Build GRCm38
- UniGene ID
- Mm.2060
- Species
- Mus musculus
- Gene Symbol
- 1700028B04Rik
- Entrez Gene ID
- 70001
- Gene Name
- NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 28496941 - 28497492 on Build GRCm38
- UniGene ID
- Mm.417851
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006445.1 |
- |
|
678 |
|
View Full Product Details