Gene Transcripts

Gene Symbol
Fundc2
Entrez Gene ID
67391
Gene Name
FUN14 domain containing 2
Gene Aliases
4833415N24Rik, DC44, HCBP6
Chromosome Location
Chr. X: 75382399 - 75397158 on Build GRCm38
UniGene ID
Mm.126404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026126.4 NP_080402.3 5 1489
GenBank mRNA AK031214.1 - 5 1466
AK076262.1 - 64

View Full Product Details