Gene Transcripts

Gene Symbol
Dph2
Entrez Gene ID
67728
Gene Name
DPH2 homolog (S. cerevisiae)
Gene Aliases
9130020C19Rik, AI467389, Dph2l2
Chromosome Location
Chr. 4: 117888643 - 117892048 on Build GRCm38
UniGene ID
Mm.13823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026344.3 NP_080620.1 6 1996
GenBank mRNA AK011199.1 - 6 2014
AK018638.1 - 6 1997
AK051026.1 - 6 2016
AK080747.1 - 4 2293
AK194858.1 - 1 105
AK218933.1 - 1 133
BC014746.1 - 5 1892
BC031124.1 - 6 1983 5029876

View Full Product Details