Gene Transcripts

Gene Symbol
Nudt22
Entrez Gene ID
68323
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 22
Gene Aliases
0610006K04Rik, AI851793, AW545547
Chromosome Location
Chr. 19: 6993019 - 6996115 on Build GRCm38
UniGene ID
Mm.49041
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026675.2 NP_080951.1 5 790
GenBank mRNA AK002252.1 - 5 791
BC019768.1 - 4 767 5124658

View Full Product Details