Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020605.3 | NP_065630.1 | 5 | 2330 | |
GenBank mRNA | AB024449.1 | - | 5 | 2326 | |
AK045706.1 | - | 5 | 2317 | ||
AK046136.1 | - | 2323 | |||
AK082880.1 | - | 5 | 2319 | ||
BC103682.1 | - | 5 | 2285 | 40047806 | |
BC104736.1 | - | 5 | 2285 | 40047803 | |
BC105307.1 | - | 5 | 2285 | 40047801 |