Gene Transcripts
- Gene Symbol
- Slc29a1
- Entrez Gene ID
- 63959
- Gene Name
- solute carrier family 29 (nucleoside transporters), member 1
- Gene Aliases
- 1200014D21Rik, AA407560, ENT1
- Chromosome Location
- Chr. 17: 45585200 - 45599608 on Build GRCm38
- UniGene ID
- Mm.29744
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199113.1 |
NP_001186042.1
|
4, 5 |
355 |
|
NM_001199114.1 |
NP_001186043.1
|
3, 4 |
194 |
|
NM_001199115.1 |
NP_001186044.1
|
3, 4 |
300 |
|
NM_001199116.1 |
NP_001186045.1
|
3, 4 |
260 |
|
NM_022880.3 |
NP_075018.1
|
2, 3 |
301 |
|
GenBank mRNA |
AF131212.1 |
- |
2, 3 |
302 |
|
AF305501.1 |
- |
3, 4 |
265 |
|
AK004756.1 |
- |
3, 4 |
246 |
|
AK050089.1 |
- |
3, 4 |
285 |
|
AK146265.1 |
- |
3, 4 |
195 |
|
AK149483.1 |
- |
3, 4 |
216 |
|
AK170463.1 |
- |
3, 4 |
232 |
|
AK177360.1 |
- |
|
294 |
|
AK178838.1 |
- |
|
128 |
|
AK180131.1 |
- |
|
292 |
|
AK180168.1 |
- |
|
190 |
|
AK183203.1 |
- |
|
291 |
|
AK191284.1 |
- |
|
293 |
|
AK197634.1 |
- |
|
292 |
|
AK205425.1 |
- |
|
291 |
|
AK205461.1 |
- |
|
292 |
|
AK206588.1 |
- |
|
191 |
|
AK218485.1 |
- |
|
182 |
|
BB853265.1 |
- |
4, 5 |
365 |
|
BC004828.1 |
- |
4, 5 |
318 |
2631728 |
BC006812.1 |
- |
2, 3 |
232 |
3155188 |
BY055687.1 |
- |
3, 4 |
195 |
|
CD540408.1 |
- |
2, 3 |
220 |
|
CX219686.1 |
- |
4, 5 |
350 |
|
EU180577.1 |
- |
2, 3 |
170 |
|
View Full Product Details