Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024190.2 | NP_077152.1 | 1 | 1475 | |
GenBank mRNA | AK012996.1 | - | 1 | 1426 | |
AK014091.1 | - | 1 | 1438 | ||
AK151071.1 | - | 1 | 1445 | ||
AK167614.1 | - | 1 | 1413 | ||
BC002229.1 | - | 1 | 1411 | 3491354 | |
BC034442.1 | - | 1 | 1399 |