Gene Transcripts

Gene Symbol
Dnajc15
Entrez Gene ID
66148
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 15
Gene Aliases
1110003P16Rik, Dnajd1
Chromosome Location
Chr. 14: 77826217 - 77874917 on Build GRCm38
UniGene ID
Mm.248046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025384.3 NP_079660.1 642
GenBank mRNA BC027509.1 - 642

View Full Product Details