Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025670.4 | NP_079946.2 | 6 | 1477 | |
GenBank mRNA | AK005157.1 | - | 6 | 1463 | |
AK017480.1 | - | 6 | 1460 | ||
AK031705.1 | - | 6 | 1479 | ||
AK035791.1 | - | 4 | 2772 | ||
AK051560.1 | - | 6 | 1475 | ||
AK149939.1 | - | 6 | 1473 | ||
AK150219.1 | - | 6 | 1422 | ||
AK166141.1 | - | 6 | 1470 | ||
BC004063.1 | - | 6 | 1476 | 3590511 |