Gene Transcripts
- Gene Symbol
- Zdhhc6
- Entrez Gene ID
- 66980
- Gene Name
- zinc finger, DHHC domain containing 6
- Gene Aliases
- 2400007G07Rik, 5930409M18Rik, AI462345, C77369
- Chromosome Location
- Chr. 19: 55298293 - 55316032 on Build GRCm38
- UniGene ID
- Mm.386789
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033573.1 |
NP_001028745.1
|
6, 7 |
1301 |
|
NM_025883.3 |
NP_080159.3
|
6, 7 |
1185 |
|
GenBank mRNA |
AK010296.1 |
- |
6, 7 |
1180 |
|
AK011373.1 |
- |
6, 7 |
1152 |
|
AK035172.1 |
- |
|
1264 |
|
AK038177.1 |
- |
|
1164 |
|
AK050388.1 |
- |
6, 7 |
1300 |
|
AK077693.1 |
- |
6, 7 |
1139 |
|
AK077838.1 |
- |
|
1150 |
|
AK139707.1 |
- |
6, 7 |
1142 |
|
AK144128.1 |
- |
6, 7 |
1157 |
|
AK154741.1 |
- |
6, 7 |
1163 |
|
AK157494.1 |
- |
6, 7 |
1167 |
|
AK159304.1 |
- |
6, 7 |
1186 |
|
AK168497.1 |
- |
7, 8 |
1308 |
|
BC033317.1 |
- |
6, 7 |
1132 |
4459242 |
View Full Product Details