Gene Transcripts

Gene Symbol
Gigyf1
Entrez Gene ID
57330
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
Perq1, mKIAA4110
Chromosome Location
Chr. 5: 137510258 - 137527935 on Build GRCm38
UniGene ID
Mm.268344
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031408.2 NP_113596.2 17 2084
GenBank mRNA AK220289.1 - 10 1093
AY176042.1 - 17 2100
BC052875.1 - 11 1188

View Full Product Details