Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018861.3 | NP_061349.3 | 3 | 827 | |
GenBank mRNA | AF246129.1 | - | 3 | 586 | |
AK044280.1 | - | 3 | 798 | ||
AK136900.1 | - | 3 | 827 | ||
AK139248.1 | - | 3 | 826 | ||
AK140866.1 | - | 3 | 828 | ||
AK148075.1 | - | 3 | 828 | ||
BC043483.1 | - | 3 | 785 | 5003744 | |
BC052733.1 | - | 3 | 701 | 6816689 | |
U75215.1 | - | 3 | 697 |