Gene Transcripts

Gene Symbol
Tnfsf14
Entrez Gene ID
50930
Gene Name
tumor necrosis factor (ligand) superfamily, member 14
Gene Aliases
HVEM-L, HVEML, LIGHT, LTg, Ly113
Chromosome Location
Chr. 17: 57189474 - 57194181 on Build GRCm38
UniGene ID
Mm.483369
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019418.3 NP_062291.1 4 1841
GenBank mRNA AB029155.1 - 4 1852
BY554892.1 - 1 393

View Full Product Details