Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019684.1 | NP_062658.1 | 5 | 459 | |
GenBank mRNA | AF043288.1 | - | 5 | 459 | |
AJ748651.1 | - | 4 | 283 | ||
AK004696.2 | - | 5 | 477 | ||
BC120883.1 | - | 5 | 454 | 40057934 | |
BC120884.1 | - | 5 | 454 | 40057937 |