Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290315.1 | NP_001277244.1 | 3 | 525 | |
NM_020043.3 | NP_064427.2 | 3 | 525 | ||
GenBank mRNA | AB052620.1 | - | 3 | 525 | |
AB052621.1 | - | 3 | 525 | ||
AF176694.1 | - | 3 | 476 | ||
AK079073.1 | - | 3 | 490 | ||
AK082162.1 | - | 3 | 519 | ||
AK122535.1 | - | 454 | |||
AK143256.1 | - | 3 | 478 | ||
BB624536.1 | - | 3 | 516 | ||
CD352400.1 | - | 2 | 162 |