Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290315.1 | NP_001277244.1 | 12 | 2205 | |
NM_020043.3 | NP_064427.2 | 12 | 2202 | ||
GenBank mRNA | AB052620.1 | - | 12 | 2202 | |
AB052621.1 | - | 12 | 2205 | ||
AF176694.1 | - | 12 | 2153 | ||
AK122535.1 | - | 2281 | |||
AK143256.1 | - | 11 | 2664 | ||
BU701299.1 | - | 2 | 195 |