Gene Transcripts

Gene Symbol
Pate4
Entrez Gene ID
56872
Gene Name
prostate and testis expressed 4
Gene Aliases
9530004K16Rik, Pate-B, Svs7
Chromosome Location
Chr. 9: 35607093 - 35616371 on Build GRCm38
UniGene ID
Mm.390143
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020264.4 NP_064660.2 2 189
GenBank mRNA AF134204.1 - 2 150
AK020540.1 - 2 173
BC120764.1 - 2 189 40129687
BC120766.1 - 2 189 40129689

View Full Product Details