Gene Transcripts

Gene Symbol
Ubfd1
Entrez Gene ID
28018
Gene Name
ubiquitin family domain containing 1
Gene Aliases
AI467302, D7Wsu105e, D7Wsu128e, Ubph
Chromosome Location
Chr. 7: 122067198 - 122082199 on Build GRCm38
UniGene ID
Mm.282122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138589.2 NP_613055.2 7 1528
GenBank mRNA AK017851.1 - 7 1302
AK140891.1 - 7 1289

View Full Product Details