Gene Transcripts
- Gene Symbol
- Tgfb1i1
- Entrez Gene ID
- 21804
- Gene Name
- transforming growth factor beta 1 induced transcript 1
- Gene Aliases
- ARA55, Hic5, TSC-5, hic-5
- Chromosome Location
- Chr. 7: 128246799 - 128255699 on Build GRCm38
- UniGene ID
- Mm.3248
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289550.1 |
NP_001276479.1
|
8, 9 |
1065 |
|
NM_001289551.1 |
NP_001276480.1
|
8, 9 |
1061 |
|
NM_001289552.1 |
NP_001276481.1
|
7, 8 |
922 |
|
NM_001289553.1 |
NP_001276482.1
|
7, 8 |
949 |
|
GenBank mRNA |
AK003170.1 |
- |
8, 9 |
899 |
|
AK041233.1 |
- |
8, 9 |
894 |
|
AK085042.1 |
- |
7, 8 |
791 |
|
AK087114.1 |
- |
8, 9 |
896 |
|
AK156423.1 |
- |
8, 9 |
900 |
|
AK158409.1 |
- |
7, 8 |
774 |
|
BC002049.1 |
- |
8, 9 |
899 |
3489459 |
BC056362.1 |
- |
7, 8 |
2174 |
6816930 |
DQ143891.1 |
- |
7, 8 |
950 |
|
DQ143892.1 |
- |
6, 7 |
736 |
|
DQ143893.1 |
- |
6, 7 |
743 |
|
DQ143894.1 |
- |
6, 7 |
738 |
|
DQ143895.1 |
- |
6, 7 |
1154 |
|
DQ143896.1 |
- |
6, 7 |
1115 |
|
DQ143897.1 |
- |
6, 7 |
1193 |
|
DQ143898.1 |
- |
6, 7 |
1142 |
|
DQ143899.1 |
- |
5, 6 |
1269 |
|
DQ143900.1 |
- |
7, 8 |
962 |
|
L22482.1 |
- |
7, 8 |
1026 |
|
View Full Product Details