Gene Transcripts

Gene Symbol
Tulp3
Entrez Gene ID
22158
Gene Name
tubby-like protein 3
Gene Aliases
2310022L06Rik, AI316887
Chromosome Location
Chr. 6: 128321161 - 128355872 on Build GRCm38
UniGene ID
Mm.291783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011657.2 NP_035787.1 4, 5 510
GenBank mRNA AB016963.1 - 4, 5 453
AF045582.1 - 4, 5 463
BC060068.1 - 4, 5 440 6419643

View Full Product Details