Gene Transcripts

Gene Symbol
Dusp13
Entrez Gene ID
27389
Gene Name
dual specificity phosphatase 13
Gene Aliases
DUSP13A, DUSP13B, Gm1203, LMW-DSP6, MDSP, TMDP, TS-DSP6
Chromosome Location
Chr. 14: 21733395 - 21748622 on Build GRCm38
UniGene ID
Mm.390674
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007268.1 NP_001007269.1 7 1109
NM_013849.3 NP_038877.2 3 434
GenBank mRNA AB027003.1 - 2 215
AF237620.1 - 3 417
AK006247.1 - 3 435
AY674052.1 - 7 1109
BC100421.1 - 2 340 30919819

View Full Product Details