Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277957.1 | NP_001264886.1 | 7 | 999 | |
NM_001277958.1 | NP_001264887.1 | 7 | 988 | ||
NM_001277959.1 | NP_001264888.1 | 7 | 967 | ||
NM_021466.3 | NP_067441.1 | 7 | 999 | ||
GenBank mRNA | AK132811.1 | - | 7 | 953 | |
AK133426.1 | - | 7 | 989 | ||
BC061106.1 | - | 7 | 884 | 6595990 | |
BC138425.1 | - | 7 | 865 | 8861445 | |
Y09972.1 | - | 7 | 901 |