Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033622.1 | NP_296371.1 | 7 | 1292 | |
GenBank mRNA | AF119383.1 | - | 7 | 1292 | |
AF352245.1 | - | 7 | 1296 | ||
AK034121.1 | - | 7 | 924 | ||
AK079180.1 | - | 7 | 1511 | ||
AK155288.1 | - | 7 | 1071 | ||
BC106840.1 | - | 6 | 1208 | 40045718 | |
BC106841.1 | - | 7 | 1265 | 40045720 |