Gene Transcripts

Gene Symbol
Galnt5
Entrez Gene ID
83627
Gene Name
polypeptide N-acetylgalactosaminyltransferase 5
Gene Aliases
Chromosome Location
Chr. 3: 44025964 - 44072073 on Build Rnor_6.0
UniGene ID
Rn.30048
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031796.1 NP_113984.1 1 300
GenBank mRNA AF049344.1 - 1 300

View Full Product Details