Gene Transcripts

Gene Symbol
Pgf
Entrez Gene ID
94203
Gene Name
placental growth factor
Gene Aliases
Plgf
Chromosome Location
Chr. 6: 108994016 - 109004598 on Build Rnor_6.0
UniGene ID
Rn.6960
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053595.2 NP_446047.1 7 839
GenBank mRNA BC087006.1 - 7 839
L40030.1 - 7 574

View Full Product Details