Gene Transcripts

Gene Symbol
Ap2m1
Entrez Gene ID
116563
Gene Name
adaptor-related protein complex 2, mu 1 subunit
Gene Aliases
Ap50, Clapm1, mu2
Chromosome Location
Chr. 11: 84041184 - 84047542 on Build Rnor_6.0
UniGene ID
Rn.3172
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053837.1 NP_446289.1 2 280
GenBank mRNA BC087724.1 - 287
M23674.1 - 2 280

View Full Product Details