Gene Transcripts
- Gene Symbol
- Ugt2b1
- Entrez Gene ID
- 286954
- Gene Name
- UDP glucuronosyltransferase 2 family, polypeptide B1
- Gene Aliases
- 2B1, Udpgtr2, Ugt2b17
- Chromosome Location
- Chr. 14: 22724399 - 22736148 on Build Rnor_6.0
- UniGene ID
- Rn.9969
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173295.1 |
NP_775417.1
|
3 |
997 |
|
GenBank mRNA |
M13506.1 |
- |
3 |
997 |
|
- Gene Symbol
- RGD1565664
- Entrez Gene ID
- 498344
- Gene Name
- similar to UDP-glucuronosyltransferase 2B1 precursor, microsomal (UDPGTR-2)
- Gene Aliases
- Chromosome Location
- Chr. 14: 22768150 - 22780691 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details