Gene Transcripts

Gene Symbol
Ltb4r
Entrez Gene ID
59264
Gene Name
leukotriene B4 receptor
Gene Aliases
Ltb4r1
Chromosome Location
Chr. 15: 34449302 - 34453352 on Build Rnor_6.0
UniGene ID
Rn.41003
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021656.1 NP_067688.1 1 536
XM_006252011.2 XP_006252073.1 2 2210
XM_006252012.1 XP_006252074.1 2 572

View Full Product Details