Gene Transcripts
- Gene Symbol
- SSR3
- Entrez Gene ID
- 6747
- Gene Name
- signal sequence receptor subunit 3
- Gene Aliases
- TRAPG
- Chromosome Location
- Chr. 3: 156539553 - 156555200 on Build GRCh38
- UniGene ID
- Hs.518346
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308204.1 |
NP_001295133.1
|
3, 4 |
329 |
|
NM_001308205.1 |
NP_001295134.1
|
3, 4 |
353 |
|
NM_007107.4 |
NP_009038.1
|
3, 4 |
464 |
|
GenBank mRNA |
AF087907.1 |
- |
3, 4 |
412 |
|
AF110647.1 |
- |
3, 4 |
409 |
|
AK312935.1 |
- |
3, 4 |
371 |
|
BC017203.1 |
- |
3, 4 |
410 |
3618589 |
BT006669.1 |
- |
3, 4 |
353 |
|
DA135338.1 |
- |
3, 4 |
448 |
|
DA283706.1 |
- |
3, 4 |
329 |
|
DA725890.1 |
- |
3, 4 |
355 |
|
DA750309.1 |
- |
3, 4 |
448 |
|
HY035536.1 |
- |
|
464 |
|
View Full Product Details