Gene Transcripts

Gene Symbol
CA5B
Entrez Gene ID
11238
Gene Name
carbonic anhydrase 5B
Gene Aliases
CA-VB
Chromosome Location
Chr. X: 15738281 - 15787625 on Build GRCh38
UniGene ID
Hs.653287
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007220.3 NP_009151.1 3 340
XM_005274442.4 XP_005274499.1 4 428
XM_017029247.1 XP_016884736.1 2 126
GenBank mRNA AB021660.1 - 3 360
AK291050.1 - 3 359
BC028142.2 - 2 277 5214534

Gene Symbol
CA5BP1
Entrez Gene ID
340591
Gene Name
carbonic anhydrase 5B pseudogene 1
Gene Aliases
CA5BL, CA5BP, PRO2325
Chromosome Location
Chr. X: 15674916 - 15703351 on Build GRCh38
UniGene ID
Hs.732244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026551.1 3 211
GenBank mRNA CD675860.1 - 3 220
DB271346.1 - 2 126

View Full Product Details