Gene Transcripts

Gene Symbol
Ddit4
Entrez Gene ID
140942
Gene Name
DNA-damage-inducible transcript 4
Gene Aliases
Rtp801
Chromosome Location
Chr. 20: 29509283 - 29511382 on Build Rnor_6.0
UniGene ID
Rn.9775
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080906.2 NP_543182.1 3 1020
GenBank mRNA AF334162.1 - 3 1025
BC062021.1 - 3 953 5623446

View Full Product Details