Gene Transcripts

Gene Symbol
Esm1
Entrez Gene ID
64536
Gene Name
endothelial cell-specific molecule 1
Gene Aliases
Pg25
Chromosome Location
Chr. 2: 45104392 - 45113130 on Build Rnor_6.0
UniGene ID
Rn.6376
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022604.2 NP_072126.1 3 970
GenBank mRNA BC070888.1 - 3 970
U80818.1 - 3 883

View Full Product Details