Gene Transcripts

Gene Symbol
Duox2
Entrez Gene ID
79107
Gene Name
dual oxidase 2
Gene Aliases
Thox2
Chromosome Location
Chr. 3: 114218187 - 114237536 on Build Rnor_6.0
UniGene ID
Rn.55542
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024141.1 NP_077055.1 9 1057
XM_006234862.2 XP_006234924.1 11 2021
GenBank mRNA AF237962.1 - 9 1057

View Full Product Details