Gene Transcripts

Gene Symbol
Cdc37
Entrez Gene ID
114562
Gene Name
cell division cycle 37
Gene Aliases
Chromosome Location
Chr. 8: 22160922 - 22173797 on Build Rnor_6.0
UniGene ID
Rn.17982
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053743.1 NP_446195.1 3 533
GenBank mRNA AB097113.1 - 3 446
BC061720.1 - 3 502 5598479
D26564.1 - 3 533

View Full Product Details