Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001324444.1 | NP_001311373.1 | 9 | 1486 | |
NM_001324445.1 | NP_001311374.1 | 8 | 2201 | ||
NM_001324446.1 | NP_001311375.1 | 9 | 2219 | ||
NM_001324452.1 | NP_001311381.1 | 7 | 2146 | ||
NM_012091.4 | NP_036223.2 | 9 | 1428 | ||
GenBank mRNA | AF125188.1 | - | 8 | 1308 | |
AK001687.1 | - | 9 | 1419 | ||
AK296834.1 | - | 10 | 1299 | ||
AK314652.1 | - | 9 | 1370 | ||
BC002758.2 | - | 9 | 1374 | 3632079 |