Gene Transcripts

Gene Symbol
Rps13
Entrez Gene ID
161477
Gene Name
ribosomal protein S13
Gene Aliases
Chromosome Location
Chr. 1: 185328902 - 185331332 on Build Rnor_6.0
UniGene ID
Rn.103229
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130432.2 NP_569116.1 3 135
GenBank mRNA AA686236.1 - 3 137
BC084724.1 - 3 111 7318693
FQ209976.1 - 3 133
FQ209999.1 - 3 132
FQ210203.1 - 3 129
FQ210309.1 - 3 128
FQ217627.1 - 3 129
FQ220591.1 - 3 127
FQ220785.1 - 3 129
FQ221555.1 - 3 129
FQ221602.1 - 3 131
FQ221859.1 - 3 132
FQ222187.1 - 3 129
FQ222199.1 - 3 129
FQ222266.1 - 3 132
FQ222333.1 - 3 129
FQ222359.1 - 3 129
FQ222425.1 - 2 80
FQ222441.1 - 3 132
FQ222573.1 - 3 129
FQ222592.1 - 3 129
FQ222643.1 - 3 129
FQ222649.1 - 3 129
FQ222885.1 - 3 129
FQ223174.1 - 3 129
FQ223351.1 - 3 129
FQ223601.1 - 3 129
FQ223978.1 - 3 129
FQ224030.1 - 3 129
FQ224170.1 - 3 148
FQ225803.1 - 3 127
FQ228324.1 - 3 126
X53378.1 - 3 130

Gene Symbol
LOC683961
Entrez Gene ID
683961
Gene Name
similar to ribosomal protein S13
Gene Aliases
Chromosome Location
Chr. 5: 57652427 - 57658436 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749925.3 XP_003749973.2 1 128

Gene Symbol
LOC684988
Entrez Gene ID
684988
Gene Name
similar to ribosomal protein S13
Gene Aliases
Chromosome Location
Chr. 3: 4341229 - 4341759 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001053043.4 XP_001053043.1 1 121

View Full Product Details