Gene Transcripts

Gene Symbol
Tnni3k
Entrez Gene ID
295531
Gene Name
TNNI3 interacting kinase
Gene Aliases
Cark
Chromosome Location
Chr. 2: 261069210 - 261337296 on Build Rnor_6.0
UniGene ID
Rn.3434
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181769.1 NP_861434.1 3 211
XM_008761563.1 XP_008759785.1 3 375
GenBank mRNA AY303692.1 - 3 211
BC098648.1 - 3 216

View Full Product Details