Gene Transcripts

Gene Symbol
Rab15
Entrez Gene ID
299156
Gene Name
RAB15, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 6: 99845952 - 99870320 on Build Rnor_6.0
UniGene ID
Rn.24234
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198749.2 NP_942044.1 1 476
GenBank mRNA BC094954.1 - 1 476
M83679.1 - 1 320

View Full Product Details