Gene Transcripts
- Gene Symbol
- Glra1
- Entrez Gene ID
- 25674
- Gene Name
- glycine receptor, alpha 1
- Gene Aliases
- GLYRA1
- Chromosome Location
- Chr. 10: 40855559 - 40953803 on Build Rnor_6.0
- UniGene ID
- Rn.10109
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013133.1 |
NP_037265.1
|
9 |
1922 |
|
GenBank mRNA |
D00833.1 |
- |
9 |
1922 |
|
View Full Product Details