Gene Transcripts

Gene Symbol
Rgs9
Entrez Gene ID
29481
Gene Name
regulator of G-protein signaling 9
Gene Aliases
Chromosome Location
Chr. 10: 97509971 - 97582188 on Build Rnor_6.0
UniGene ID
Rn.10903
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019224.2 NP_062097.1 2 226
GenBank mRNA AB019145.1 - 2 226
AF071475.1 - 2 222

View Full Product Details